missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate, 96%

Product Code. 10290060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10290060 1 g Glass bottle 1g
10516830 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10290060 Supplier Thermo Scientific Acros Supplier No. 427670010
 Restricted Item
Estimated Shipment: 09-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 129946-88-9 | C14H8F6O3S2 | 402.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 129946-88-9
Molecular Formula C14H8F6O3S2
Molecular Weight (g/mol) 402.33
MDL Number MFCD00236132
InChI Key QXXHXTRTGZBOGD-UHFFFAOYSA-M
Synonym 5-trifluoromethyl-5h-dibenzo b,d thiophen-5-ium trifluoromethanesulfonate, s-trifluoromethyl dibenzothiophenium trifluoromethanesulfonate, 5-trifluoromethyl dibenzothiophenium trifluoromethanesulfonate, s-trifluoromethyl dibenzothiophenium triflate, s-trifluoromethyl dibenzothiophenium trifluoromethanesulphonate, trifluoromethanesulfonate; 5-trifluoromethyl dibenzothiophen-5-ium, pubchem16425, acmc-20a6nt, c13h8f3s.chf3o3s, s-trifluoromethyl dibenzothiophenium trifluoromet
PubChem CID 2777507
IUPAC Name trifluoromethanesulfonate;5-(trifluoromethyl)dibenzothiophen-5-ium
SMILES [O-]S(=O)(=O)C(F)(F)F.FC(F)(F)[S+]1C2=CC=CC=C2C2=CC=CC=C12

Spezifikation

CAS Min % 95.0
CAS Max % 100.0
Melting Point 150°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 402.33
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.