missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)indole, 98%

Product Code. 11482140
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482140 500 mg 500mg
11472140 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482140 Supplier Thermo Scientific Alfa Aesar Supplier No. H28782.ME
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100846-24-0 | C9H6F3N | 185.149 g/mol

In 3-Naphthylindole construction by rhodium (II)-catalyzed regioselective direct arylation of indoles with 1-diazonaphthalen-2-(1H)-ones treatment of 5-(trifluoromethyl)indole(1k) or 6-(trifluoromethyl)indole (1l) with 2a afforded the desired products 3k (86%)and 3l (86%).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In 3-Naphthylindole construction by rhodium (II)-catalyzed regioselective direct arylation of indoles with 1-diazonaphthalen-2-(1H)-ones treatment of 5-(trifluoromethyl)indole(1k) or 6-(trifluoromethyl)indole (1l) with 2a afforded the desired products 3k (86%)and 3l (86%).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100846-24-0
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.149
MDL Number MFCD03095341
InChI Key LCFDJWUYKUPBJM-UHFFFAOYSA-N
Synonym 5-trifluoromethyl indole, 5-trifluoromethyl-1h-indole, 5-trifluoromethylindole, 1h-indole, 5-trifluoromethyl, zlchem 948, pubchem1712, acmc-1box4, 5-trifluoromethyl-indole
PubChem CID 2763075
IUPAC Name 5-(trifluoromethyl)-1H-indole
SMILES C1=CC2=C(C=CN2)C=C1C(F)(F)F

Specifications

Melting Point 68°C to 71°C
Quantity 500 mg
Formula Weight 185.15
Percent Purity 98%
Chemical Name or Material 5-(Trifluoromethyl)indole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.