missing translation for 'onlineSavingsMsg'
Learn More

(5-Trifluoromethylpyridin-3-yl)boronic acid, 95%

Product Code. 10687220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10687220 250 mg Glass bottle 250mg
10506260 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10687220 Supplier Thermo Scientific Acros Supplier No. 430522500
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 947533-51-9 | C6H5BF3NO2 | 190.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947533-51-9
Molecular Formula C6H5BF3NO2
Molecular Weight (g/mol) 190.92
MDL Number MFCD09952041
InChI Key SFBQNNGMEKUJAN-UHFFFAOYSA-N
Synonym 5-trifluoromethyl-pyridine-3-boronic acid, 5-trifluoromethyl pyridin-3-yl boronic acid, 3-trifluoromethylpyridine-5-boronic acid, 3-trifluoromethyl-5-pyridyl boronic acid, 5-trifluoromethyl pyridine-3-boronic acid, 5-trifluoromethyl pyridin-3-ylboronic acid, 5-trifluoromethylpyridine-3-boronic acid, 5-trifluoromethyl-3-pyridyl boronic acid, 3-borono-5-trifluoromethyl pyridine
PubChem CID 45158900
IUPAC Name [5-(trifluoromethyl)pyridin-3-yl]boronic acid
SMILES OB(O)C1=CC(=CN=C1)C(F)(F)F

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 260.0°C to 262.6°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 190.92
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material (5-Trifluoromethylpyridin-3-yl)boronic acid
Show More Show Less
compliance-icons
Product Identifier
  • (5-Trifluoromethylpyridin-3-yl)boronic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.