missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ STO-609 acetate

Description
STO-609 acetate is a selective, cell-permeable inhibitor of Ca2+-calmodulin-dependent protein kinase kinase (Ki values are 80 and 15 ng/ml for inhibition of CaM-KKα and CaM-KKβ respectively); competes for the ATP-binding site. Displays > 80-fold selectivity over CaMK1, CaMK2, CaMK4, MLCK, PKC, PKA and p42 MAPK.
Specifications
Specifications
| Chemical Name or Material | 7-Oxo-7H-benzimidazo[2,1-a]benz[de]isoquinoline-3-carboxylic acid acetate |
| CAS | 1173022-21-3 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Solubility | Soluble to 10mM with sonication in DMSO |
| Molecular Formula | C19H10N2O3.C2H4O2 |
| Formula Weight | Observed MW: 374.35 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?