missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VX 765

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
VX 765 is a high affinity and selective caspase-4 and caspase 1/ICE inhibitor (Ki of <0.6 and 0.8 nM, respectively). Prodrug of VRT-043198. Inhibits LPS-induced IL-1β production ∼ 60% in collagen-induced arthritis mouse model. Exhibits anticonvulsant effects in mouse model of acute seizures. Anti-inflammatory. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N-(4-Amino-3-chlorobenzoyl)-3-methyl-L-valyl-N-[(2 R,3 S)-2-ethoxytetrahydro-5-oxo-3-furanyl]-L-prolinamide |
| CAS | 273404-37-8 |
| Quantity | 50 mg |
| Target | Caspase Inhibitors |
| Molecular Formula | C24H33ClN4O6 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction