missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Turbidity Reference Standard, Turbidity Reference Material.
- Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
- Certified
- NIST traceable
- Non-toxic, non-carcinogenic
- Highly accurate
- Stable
- EPA approved
Specifications
Specifications
| For Use With (Equipment) | Ratio Instruments |
| Description | Liquid format in HDPE Bottle GL45 |
| Accuracy | Accuracy ± 1% NTU |
| Turbidity | 50 NTU ± 1% |
| Volume (Metric) | 500 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
