missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek 50 TS Microplate Washer
The BioTek 50 TS washer offers excellent microplate washing performance. For Research Use Only. Not for use in diagnostic procedures.
Brand: Agilent BioTek 50TS12V-SN
Description
The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well
The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.
Services
12 months
Specifications
CE and TUV marked. RoHS compliant. | |
Optional (2 to 4 L) | |
Programmable in minutes and seconds, up to 30 min. | |
9.8 kg | |
25 to 3000 μL/well | |
150 to 1000 μL/well/sec. (manifold dependent) | |
50TS12V | |
4.3 in. Color LCD Touch Screen Display |
USB (1) | |
24-, 96-, 384-well plates (96- and 384- well: standard and low profile plates; solid and filter bottom (option), 24-well plates with 4-well manifold) | |
Microplate Washer | |
1 to 10 | |
4.3 in. LCD Color Touch Screen | |
One positive displacement syringe drive | |
96-well plates only; Buffer Switching | |
22 lb. |