missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 50 TS Microplate Washer

The BioTek 50 TS washer offers excellent microplate washing performance. For Research Use Only. Not for use in diagnostic procedures.

Brand:  Agilent BioTek 50TS16-SN


Additional Details : Weight : 10.00000kg

 View more versions of this product

Product Code. 17227789

  • POA / Each
Estimated Shipment: 13-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »


Description

Description

The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well

The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.

Services

12 months

Specifications

Specifications

CE and TUV marked. RoHS compliant.
Optional (2 to 4 L)
Programmable in minutes and seconds, up to 30 min.
9.8 kg
25 to 3000 μL/well
150 to 1000 μL/well/sec. (manifold dependent)
50TS16
4.3 in. Color LCD Touch Screen Display
USB (1)
24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold)
Microplate Washer
1 to 10
4.3 in. LCD Color Touch Screen
One positive displacement syringe drive
96-/384-well plates
22 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers