missing translation for 'onlineSavingsMsg'
Learn More

Jenway™ 500μl glass cuvette (min 320nm - 4 windows with PTFE lid)

Product Code. 11484919
Change view
Click to view available options
Capacity (Metric):
500 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11484919 500 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11484919 Supplier Jenway™ Supplier No. 6800617
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Jenway Fluorimeters

  • For use with Jenway Fluorimeters
  • 500μl glass cuvette
  • min 320nm
  • 4 windows with PTFE lid

Specifications

Description Minimum 320nm, 4 windows with PTFE lid
Capacity (Metric) 500 μL
Material Glass
Wavelength Range 360 to 2500 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.