Learn More
Thermo Scientific™ 500μL PTFE Luer-Lok, Gas-Tight Syringe
Product Code. 15214593 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
500 μL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
15214593 500 μL Each
1 options
Product Code. 15214593 Supplier Thermo Scientific™ Supplier No. 365J7659
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Thermo Scientific Luer Syringes offer easy, accurate and reproducible injections for a variety of volumes.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gas Tight GC Autosampler Syringe, PTFE Luer-Lok
Volume (Metric) 500 μL
For Use With (Equipment) Gas Chromatography Systems