missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 5000 Series 0.03μm to 0.30μm Polymer Particle Suspensions

Product Code. 11829812 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
0.03 μm
0.06 μm
0.08 μm
0.09 μm
0.10 μm
0.11 μm
0.12 μm
0.14 μm
0.16 μm
0.17 μm
0.20 μm
0.22 μm
0.24 μm
0.26 μm
0.3 μm
Quantity:
15 mL
100 mL
1000 mL
Unit Size:
1000mL
100mL
15mL
45 product options available for selection
Product selection table with 45 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Quantity Unit Size
11829812 0.14 μm 15 mL 15mL
10184734 0.03 μm 15 mL 15mL
11829792 0.03 μm 100 mL 100mL
11839792 0.03 μm 1000 mL 1000mL
11849792 0.06 μm 15 mL 15mL
11859792 0.06 μm 100 mL 100mL
11869792 0.06 μm 1000 mL 1000mL
11879792 0.08 μm 15 mL 15mL
11889792 0.08 μm 100 mL 100mL
11899792 0.08 μm 1000 mL 1000mL
11809802 0.09 μm 15 mL 15mL
11819802 0.09 μm 100 mL 100mL
11829802 0.09 μm 1000 mL 1000mL
11839802 0.10 μm 15 mL 15mL
11849802 0.10 μm 100 mL 100mL
11859802 0.10 μm 1000 mL 1000mL
11869802 0.11 μm 15 mL 15mL
11879802 0.11 μm 100 mL 100mL
11889802 0.11 μm 1000 mL 1000mL
11899802 0.12 μm 15 mL 15mL
11809812 0.12 μm 100 mL 100mL
11819812 0.12 μm 1000 mL 1000mL
11839812 0.14 μm 100 mL 100mL
11849812 0.14 μm 1000 mL 1000mL
11859812 0.16 μm 15 mL 15mL
11869812 0.16 μm 100 mL 100mL
11879812 0.16 μm 1000 mL 1000mL
11889812 0.17 μm 15 mL 15mL
11899812 0.17 μm 100 mL 100mL
11809822 0.17 μm 1000 mL 1000mL
11819822 0.20 μm 15 mL 15mL
11829822 0.20 μm 100 mL 100mL
11839822 0.20 μm 1000 mL 1000mL
11849822 0.22 μm 15 mL 15mL
11859822 0.22 μm 100 mL 100mL
11869822 0.22 μm 1000 mL 1000mL
11598552 0.24 μm 15 mL 15mL
11889822 0.24 μm 100 mL 100mL
11899822 0.24 μm 1000 mL 1000mL
11809832 0.26 μm 15 mL 15mL
11819832 0.26 μm 100 mL 100mL
11829832 0.26 μm 1000 mL 1000mL
11839832 0.3 μm 15 mL 15mL
11849832 0.3 μm 100 mL 100mL
11859832 0.3 μm 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 45 options available.
45 options
This item is not returnable. View return policy
Product Code. 11829812 Supplier Thermo Scientific™ Supplier No. 5014A
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optimize research in light scattering, microporous filter checking, and aerosol particle generation with these particle suspensions.

Optimize research in light scattering, microporous filter checking, and aerosol particle generation with Thermo Scientific™ 5000 Series 0.03μm to 0.30μm Polymer Particle Suspensions. These particles are designed to meet the need for particulate materials with a variety of particle sizes and properties.

The polystyrene microspheres have a density of 1.05g/cm3 and a refractive index of 1.59 at 589nm. Particle diameters are measured by optical microscopy, photon correlation spectroscopy or light scattering. They are packaged as aqueous suspensions at 10% solids by weight. The 5000 Series particles range up to 3.1 microns in size. If you require larger particles, please refer to our 7000 Series Copolymer Microsphere Suspensions.

Ordering Information:

15mL, 100mL, and 1000mL bottle sizes available. 15mL (“A” Part No. bottles), 100mL (B bottles) and 1000mL (C bottles) are packaged to order; example (Part No. 5003B and 5003C). Please allow 10 working days when ordering these fill volumes.

Includes:

Material Safety Data Sheet (MSDS) and general product handling insert sheet

Recommended for:

High concentration polymer particles < 3μm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications.

TRUSTED_SUSTAINABILITY

Specifications

Additive Contains trace amount of surfactant to inhibit agglomeration and promote stability
Concentration 10%
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Description Particles useful for such applications as filter evaluation, checking and testing, light scattering research, fluid mechanics research, aerosol particle generation, dispersion studies, and many other research and development projects. They are not intended for use in instrument calibration.
Color White or Colorless
For Use With (Application) Aerosol Particle Generation, Dispersion, Filter Evaluation, Light Scattering, Particle Generation
Material Polystyrene
Product Type Polymer Particle Suspensions
Includes General product handling insert sheet
Packaging 15 mL
Particle Density 1.05g/cm3
Quantity 15 mL
Refractive Index 1.59 at 589nm (25°C)
Stability Product stable for a minimum of 24 months
Diameter (Metric) 0.14 μm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.