missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 2 and 4 12-14 kD MWCO Standard RC Dry Dialysis Kits

Product Code. 15320772
Change view
Click to view available options
Diameter (Metric):
20.4 mm
48 mm
Length (Metric):
18 mL/cm vol./length
3.3 mL/cm vol./length
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Unit Size:
1m
5m
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Quantity unitSize
15320772 48 mm 18 mL/cm vol./length 5 m Tubing/Kit 5m
15330772 48 mm 18 mL/cm vol./length 1 m Tubing/Kit 1m
15300772 20.4 mm 3.3 mL/cm vol./length 1 m Tubing/Kit 1m
15390762 20.4 mm 3.3 mL/cm vol./length 5 m Tubing/Kit 5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15320772 Supplier Spectrum™ Supplier No. 132709T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • 1m or 5m of Spectra/Por 2 or 4 dialysis tubing
  • One Standard Spectra/Por Closure (for buoyance)
  • One weighted Spectra/Por Closure (for orientation)
  • Two adhesive tubing openers

For smaller dialysis experiments or evaluating dialysis tubing for dialysis application

  • Hydrophilic, symmetric porosity
  • Ensures higher permeability and basic dialysis
  • Available as dry with glycerin (soak in water)

Specifications

MWCO 12 to 14 kDa
Diameter (Metric) 48 mm
Length (Metric) 18 mL/cm vol./length
Quantity 5 m Tubing/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.