missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 2 and 4 12-14 kD MWCO Standard RC Dry Dialysis Kits
Description
Includes
- 1m or 5m of Spectra/Por 2 or 4 dialysis tubing
- One Standard Spectra/Por Closure (for buoyance)
- One weighted Spectra/Por Closure (for orientation)
- Two adhesive tubing openers
- Hydrophilic, symmetric porosity
- Ensures higher permeability and basic dialysis
- Available as dry with glycerin (soak in water)
Specifications
Specifications
| MWCO | 12 to 14 kDa |
| Diameter (Metric) | 20.4 mm |
| Length (Metric) | 3.3 mL/cm vol./length |
| Quantity | 5 m Tubing/Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?