missing translation for 'onlineSavingsMsg'
Learn More

Lovibond Tintometer 500mL Liquid reference standard Gardner colour 8

Brand:  Lovibond Tintometer 134220


500mL Liquid reference standard Gardner colour 8

Product Code. 13505340

  • ONLINE EXCLUSIVE PRICE
    240.00 €
    233.08 € / 500mL
    Save 6.92 €
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Liquid reference standard Gardner colour 8

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1B

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P331-Do NOT induce vomiting.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : White mineral oil (petroleum), Base oil,xylene, ethylbenzene

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.