missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 2 12-14 kD MWCO Standard RC Dry Dialysis Kits

With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated

Brand:  Spectrum™ 132678T

Code : NEW

Additional Details : Weight : 20.00000kg

 View more versions of this product

Product Code. 12771138

  • 145.00 € / Each
Estimated Shipment: 11-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes:
  • 1m or 5m of Spectra/Por 2 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers

Description

Description

  • For higher permeability applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)

Specifications

Specifications

Trial Kit
Regenerated Cellulose
0.6 in.
5 m
Autoclavable
Each
12 to 14 kDa
16 mm
16 ft.
2 mL
5m roll of Spectra/Por™ 1, 2 or 3 dialysis tubing, 1 standard spectra/por closure (aids buoyancy), 1 weighted spectra/por closure (aids vertical orientation), 5 opening picks
Ideal for small volume dialysis or evaluation of membrane for an application
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers