Learn More
Cellpath System II Hex™ Tissue Processing Cassettes (Hoppered)
Product Code. 30226229
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 1500
Product Code. Color Unit Size
30226229 White Pack of 1500
1 options
Product Code. 30226229 Supplier Cellpath Supplier No. EAD010204H
  • ONLINE EXCLUSIVE PRICE
    242.37 €
    102.52 € / Pack of 1500
    Save 139.85 €
    57% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Pre-loaded into hoppers for use with automated and carousel type printers. CellPath System II Hex Tissue Processing Cassettes (Hoppered) feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
  • Pre-loaded into hoppers for use with automated and carousel type cassette printers
  • Manufactured from high–grade virgin acetyl resin
  • Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
  • Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
  • Extra thick side walls increases rigidity and stability for high quality sectioning
  • Designed to suit Thermo Printmate and Micro-writer cassette–writing machines
  • Available in 15 stock colors which color code different types of workload or applications
Specifications
Quantity 1500/Pk.
Color White
Material Acetyl Resin
Product Type Tissue Processing Cassette in Hopper tube