missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Asparagine Monohydrate, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | DL-Asparagine hydrate |
| CAS | 69833-18-7 |
| Molecular Formula | C4 H8 N2 O3 . H2 O |
| Quantity | 50 g |
| Synonym | DL-Asparagine hydrate, Asparagine, hydrate (1:1), Asparagine, monohydrate (9CI), DL-Asparagine, monohydrate |
| InChI Formula | InChI=1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2 |
| SMILES | O.NC(CC(=O)N)C(=O)O |
| IUPAC Name | 2,4-diamino-4-oxobutanoic acid;hydrate |
| Molecular Weight (g/mol) | 150.13 |
| Formula Weight | 150.0641 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction