missing translation for 'onlineSavingsMsg'
Learn More

3-Demethyl Colchicine, TRC

Product Code. 30011330
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011330 50 mg 50mg
30005733 100 mg 100mg
30003287 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011330 Supplier Toronto Research Chemicals Supplier No. TRCD23088550MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7336-33-6
Molecular Formula C21 H23 N O6
Molecular Weight (g/mol) 385.41
Synonym N-[(7S,12aRa)-3-hydroxy-1,2,10-trimethoxy-9-oxo-5,6,7,9-tetrahydrobenzo[a]heptalen-7-yl]acetamide, N-[(7S)-5,6,7,9-Tetrahydro-3-hydroxy-1,2,10-trimethoxy-9-oxobenzo[a]heptalen-7-yl]-acetamide, Acetamide, N-(5,6,7,9-tetrahydro-3-hydroxy-1,2,10-trimethoxy-9-oxobenzo[a]heptalen-7-yl)-, (S)-, Colchicine, 2-demethyl- (6CI,7CI), Colchicine, O3-demethyl- (8CI), Benzo[a]heptalene, acetamide deriv., 3-Demethyl-(-)-colchicine, 3-Demethylcolchicine, 3-Desmethylcolchicine, O3-Demethylcolchicine
IUPAC Name N-[(7S)-3-hydroxy-1,2,10-trimethoxy-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
SMILES COC1=CC=C2C(=CC1=O)[C@H](CCc3cc(O)c(OC)c(OC)c23)NC(=O)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H23NO6/c1-11(23)22-15-7-5-12-9-17(25)20(27-3)21(28-4)19(12)13-6-8-18(26-2)16(24)10-14(13)15/h6,8-10,15,25H,5,7H2,1-4H3,(H,22,23)/t15-/m0/s1
Formula Weight 385.1525
Percent Purity >95%
Chemical Name or Material 3-Demethyl Colchicine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.