missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Androsten-17beta-ol-3,6-dione, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 6-Ketotestosterone |
| CAS | 570-94-5 |
| Molecular Formula | C19 H26 O3 |
| Quantity | 50 mg |
| Synonym | Androst-4-ene-3,6-dione, 17-hydroxy-, (17β)-, Androst-4-ene-3,6-dione, 17β-hydroxy- (8CI), (17β)-17-Hydroxyandrost-4-ene-3,6-dione, 17β-Hydroxyandrost-4-ene-3,6-dione, 4-Androsten-17β-ol-3,6-dione, 6-Ketotestosterone |
| InChI Formula | InChI=1S/C19H26O3/c1-18-7-5-11(20)9-15(18)16(21)10-12-13-3-4-17(22)19(13,2)8-6-14(12)18/h9,12-14,17,22H,3-8,10H2,1-2H3/t12-,13-,14-,17-,18+,19-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CC(=O)C4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O |
| IUPAC Name | (8R,9S,10R,13S,14S,17S)-17-hydroxy-10,13-dimethyl-2,7,8,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthrene-3,6-dione |
| Molecular Weight (g/mol) | 302.41 |
| Formula Weight | 302.1882 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction