missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aceclofenac Benzyl Ester, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | benzyl[([2-[(2,6-dichlorophenyl)amino]phenyl)acetyl]oxy]acetate |
| CAS | 100499-89-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C23 H19 Cl2 N O4 |
| Quantity | 50 mg |
| Synonym | Benzyl [[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetate, Aceclofenac Imp. F (EP), Benzyl Ester of Aceclofenac |
| InChI Formula | InChI=1S/C23H19Cl2NO4/c24-18-10-6-11-19(25)23(18)26-20-12-5-4-9-17(20)13-21(27)30-15-22(28)29-14-16-7-2-1-3-8-16/h1-12,26H,13-15H2 |
| SMILES | Clc1cccc(Cl)c1Nc2ccccc2CC(=O)OCC(=O)OCc3ccccc3 |
| IUPAC Name | (2-oxo-2-phenylmethoxyethyl) 2-[2-(2,6-dichloroanilino)phenyl]acetate |
| Molecular Weight (g/mol) | 444.31 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction