missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BAM 15

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
BAM 15 is a mitochondrial protonophore uncoupler; uncouples oxidative phosphorylation in L6 myoblast mitochondria in vitro (EC50 = 270 nM). Does not depolarize the plasma membrane. Protects mice from acute renal ischemic-reperfusion injury.
Specifications
Specifications
| Chemical Name or Material | N5,N6-bis(2-Fluorophenyl)-[1,2,5]oxadiazolo[3,4-b]pyrazine-5,6-diamine |
| CAS | 210302-17-3 |
| Quantity | 50 mg |
| Target | Oxidative Phosphorylation Inhibitors |
| Molecular Formula | C16H10F2N6O |
| Formula Weight | 340.29 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction