missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bicuculline, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | (+)-Bicuculline |
| CAS | 485-49-4 |
| Molecular Formula | C20 H17 N O6 |
| Quantity | 50 mg |
| Synonym | Furo[3,4-e]-1,3-benzodioxol-8(6H)-one, 6-[(5S)-5,6,7,8-tetrahydro-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl]-, (6R)-Bicuculline (6CI,7CI,8CI), Furo[3,4-e]-1,3-benzodioxol-8(6H)-one, 6-(5,6,7,8-tetrahydro-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl)-, [R-(R*,S*)]-, 1,3-Dioxolo[4,5-g]isoquinoline, furo[3,4-e]-1,3-benzodioxol-8(6H)-one deriv., (+)-Bicuculline, Bicucullin, Bucuculline, NSC 32192, d-Bicuculline |
| InChI Formula | InChI=1S/C20H17NO6/c1-21-5-4-10-6-14-15(25-8-24-14)7-12(10)17(21)18-11-2-3-13-19(26-9-23-13)16(11)20(22)27-18/h2-3,6-7,17-18H,4-5,8-9H2,1H3/t17-,18+/m0/s1 |
| SMILES | CN1CCc2cc3OCOc3cc2[C@H]1[C@@H]4OC(=O)c5c6OCOc6ccc45 |
| IUPAC Name | (6R)-6-[(5S)-6-methyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]-6H-furo[3,4-e][1,3]benzodioxol-8-one |
| Molecular Weight (g/mol) | 367.35 |
| Formula Weight | 367.1056 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction