missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ESI 1
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ESI 1 (epigenetic silencing inhibitor 1; also called PT3) is an HDAC3 inhibitor (IC50 = 250 nM). ESI 1 exhibits high selectivity for HDAC3 over HDAC1, HDAC6, and HDAC8. Also, ESI 1 promotes remyelination in demyelinated animal models and enables de novo myelinogenesis on regenerated CNS axons. In vivo, ESI 1 enhances long-term memory in mouse model of novel object recognition. ESI 1 triggers nuclear condensate formation of lipid-metabolic regulators SREBP1/2, concentrating transcriptional co-activators to drive lipid/cholesterol biosynthesis. ESI 1 is orally bioavailable and blood-brain-barrier penetrant.
Specifications
Specifications
| Chemical Name or Material | N-(2-Aminophenyl)-5-[(phenylmethyl)amino]-2-pyrazinecarboxamide dihydrochloride |
| Quantity | 50 mg |
| Target | Class I Histone Deacetylases Inhibitors |
| Molecular Formula | C18H17N5O.2HCl |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction