missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ G 28UCM

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
G 28UCM is a FASN inhibitor. Inhibits breast cancer cell proliferation in vitro (IC50 values are 21 - 79 μM), including cell lines resistant to anti-HER2 therapies. Blocks HER2 signaling and induces apoptosis. Inhibits growth of breast cancer xenografts in mice without causing anorexia.
Specifications
Specifications
| Chemical Name or Material | 3-(3,4,5-Trihydroxybenzoyloxy)naphthalen-1-yl 3,4,5-trihydroxybenzoate |
| CAS | 1094451-90-7 |
| Quantity | 50 mg |
| Target | Fatty Acid Synthase Inhibitors |
| Molecular Formula | C24H16O10 |
| Formula Weight | 464.38 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction