missing translation for 'onlineSavingsMsg'
Learn More

L-Valyl-L-phenylalanine, MedChemExpress

Product Code. 16440477
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16440477

Brand: Medchem Express HY107378

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-Valyl-L-phenylalanine (Valylphenylalanine; H-VAL-PHE-OH) has been reported as biocompatible polymer.

L-Valyl-L-phenylalanine (Valylphenylalanine

Specifications

Chemical Name or Material L-Valyl-L-phenylalanine
Color White
CAS 3918-92-1
Physical Form Powder
Purity Grade Notes Research
Molecular Formula C14H20N2O3
Synonym Valylphenylalanine H-VAL-PHE-OH
Solubility Information DMSO : 100 mg/mL (378.33 mM; Need ultrasonic) ∣H2O : 66.67 mg/mL (252.23 mM; ultrasonic and adjust pH to 4 with HCl)
Quantity 50 mg
SMILES O=C([C@@H](N)C(C)C)N[C@H](C(O)=O)CC1=CC=CC=C1
Molecular Weight (g/mol) 264.32
Formula Weight 264.32
Percent Purity 98.95%
For Use With (Application) COVID-19-immunoregulation
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.