missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Nepicastat hydrochloride
GREENER_CHOICE

Product Code. 18744068
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18744068

Brand: Tocris Bioscience™ 5037/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective dopamine β-hydroxylase (DBH) inhibitor; orally bioavailable

Nepicastat hydrochloride is a potent and selective dopamine β-hydroxylase (DBH) inhibitor (IC50 = 9 nM) . Selective for DBH over 12 other enzymes. Decreases noradrenaline and increases dopamine levels in tissues and plasma in hypertensive rats. Prevents progressive left ventricular dysfunction in an in vivo heart failure model. Also potentiates the stimulus effects of cocaine without producing cocaine-like effects in rats. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(Aminomethyl)-1-[(2S)-5,7-difluoro-1,2,3,4-tetrahydro-2-naphthalenyl]-1,3-dihydro-2H-imidazole-2-thione hydrochloride
CAS 170151-24-3
Quantity 50 mg
Target Hydroxylase Inhibitors
Molecular Formula C14H15F2N3S.HCl
Formula Weight 331.81
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.