missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olmesartan medoxomil, MedChemExpress
Description
Olmesartan medoxomil is a potent and selective angiotensin AT1 receptor inhibitor with IC50 of 66.2 μM.
Specifications
Specifications
| Chemical Name or Material | Olmesartan medoxomil |
| CAS | 144689-63-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H30N6O6 |
| Quantity | 50 mg |
| Synonym | CS 866 |
| Solubility Information | DMSO : 50 mg/mL (89.51 mM; Need ultrasonic) |
| SMILES | O=C(OCC1=C(OC(O1)=O)C)C2=C(N=C(N2CC3=CC=C(C=C3)C4=CC=CC=C4C5=NN=NN5)CCC)C(O)(C)C |
| Molecular Weight (g/mol) | 558.59 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?