missing translation for 'onlineSavingsMsg'
Learn More

Oxiracetam, MedChemExpress

Product Code. 16424627
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16424627

Brand: Medchem Express HYB1715

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oxiracetam is a cyclic derivative of γ-aminobutyric acid (GABA) which has been commonly used as nootropic drug to treat cognitive impairments.

Oxiracetam is a cyclic derivative of γ-aminobutyric acid (GABA) which has been commonly used as nootropic drug to treat cognitive impairments.

Specifications

Chemical Name or Material Oxiracetam
CAS 62613-82-5
Color White
Purity Grade Notes Research
Molecular Formula C6H10N2O3
Quantity 50 mg
Synonym ISF2522
Solubility Information DMSO : 10 mg/mL (63.23 mM; Need ultrasonic and warming)
SMILES O=C(N)CN1C(CC(O)C1)=O
Molecular Weight (g/mol) 158.16
Formula Weight 158.16
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.