missing translation for 'onlineSavingsMsg'
Learn More

Padimate O, TRC

Product Code. 30008819
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008819 50 mL 50mL
30006258 10 mL 10mL
30010722 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008819 Supplier Toronto Research Chemicals Supplier No. TRCP13380050ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21245-02-3
Molecular Formula C17 H27 N O2
Molecular Weight (g/mol) 277.4
Synonym Benzoic acid, p-(dimethylamino)-, 2-ethylhexyl ester (8CI), 2-Ethylhexyl 4-(N,N-dimethylamino)benzoate, 2-Ethylhexyl 4-(dimethylamino)benzoate, 2-Ethylhexyl N,N-dimethyl-p-aminobenzoate, 2-Ethylhexyl p-(N,N-dimethylamino)benzoate, 2-Ethylhexyl p-(dimethylamino)benzoate, 4-(Dimethylamino)benzoic acid 2-ethylhexyl ester, Arlatone UVB, Chemcure EHA, Chivacure OPD, EHDAB, Esacure EHA, Escalol 507, Eusolex 6007, Genocure EHA, ODPABA, Octyl dimethyl PABA, Omnirad EHA, Padimate O, Quantacure EHA, Speedcure EHA
IUPAC Name 2-ethylhexyl 4-(dimethylamino)benzoate
SMILES CCCCC(CC)COC(=O)c1ccc(cc1)N(C)C

Specifications

Quantity 50 mL
InChI Formula InChI=1S/C17H27NO2/c1-5-7-8-14(6-2)13-20-17(19)15-9-11-16(12-10-15)18(3)4/h9-12,14H,5-8,13H2,1-4H3
Formula Weight 277.2042
Chemical Name or Material Padimate O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.