missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ 50SC Pump Head

Product Code. 16681444 Shop All Gilson Products
Change view
Click to view available options
Flow Rate:
50 mL/min
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
16681444 50 mL/min Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16681444 Supplier Gilson™ Supplier No. 360524
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The 50SC Pump Head is compatible with Gilson™ 305 and 306 HPLC pumping systems.

  • Compatible with Gilson™ 305 and 306 HPLC pumping systems
  • Made of stainless steel
  • Designed for quick and easy maintenance

Specifications

Flow Rate 50 mL/min
For Use With 305/306 HPLC Pumping Systems
Material Stainless Steel
Sterility Non-sterile
Type Pump Head
Maximum Pressure 2030 PSI
Case Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.