missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5alpha-Cholest-7-en-3beta-ol-1,2,5,6alpha-d4, CDN
Click to view available options
Quantity:
0.005 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Lathosterol-1,2,5,6 alpha-D4 |
| CAS | 1182005-25-9 |
| Molecular Formula | C27 D4 H42 O |
| Quantity | 0.005 g |
| Synonym | Cholest-7-en-1,2,5,6-d4-3-ol, (3β,5α)- (ACI), (3β,5α)-Cholest-7-en-1,2,5,6-d4-3-ol (ACI), 1,2,5α,6α-2H4-5α-Cholest-7-en-3β-ol, 5α-Cholest-7-en-3β-ol-1,2,5,6α-d4, Lathosterol-d4, Lathosterol-1,2,5,6α-D4 |
| InChI Formula | InChI=1 S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h10,18-21,23-25,28 H,6-9,11-17H2,1-5H3/t19-,20+,21+,23-,24+,25+,26+,27-/m1/s1/i9D,13 D,15 D,20 D/t9-,13?,15?,19+,20-,21-,23+,24-,25-,26-,27+/m0 |
| SMILES | [2 H]C1[C@H](O)C[C@]2([2 H])[C@@H]([2 H])C=C3[C@@H]4 CC[C@H]([C@H](C)CCCC(C)C)[C@@]4(C)CC[C@@H]3 [C@@]2(C)C1[2 H] |
| IUPAC Name | (3 S,5 S,6 S,9 R,10 S,13 R,14 R,17 R)-1,2,5,6-tetradeuterio-10,13-dimethyl-17-[(2 R)-6-methylheptan-2-yl]-1,2,3,4,6,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 390.6782 |
| Formula Weight | 390.38 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction