missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5alpha-Cholestane-2,2,4,4-d4, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | alpha-Cholestane-d4 |
| CAS | 205529-74-4 |
| Molecular Formula | C27 D4 H44 |
| Quantity | 0.05 g |
| Synonym | 5α-Cholestane-2,2,4,4-D4, (5α)-Cholestane-2,2,4,4-d4, α-Cholestane-d4 |
| InChI Formula | InChI=1 S/C27H48/c1-19(2)9-8-10-20(3)23-14-15-24-22-13-12-21-11-6-7-17-26(21,4)25(22)16-18-27(23,24)5/h19-25 H,6-18H2,1-5H3/t20-,21-,22+,23-,24+,25+,26+,27-/m1/s1/i7D2,11D2 |
| SMILES | [2 H]C1([2 H])CC([2 H])([2 H])[C@@H]2 CC[C@H]3 [C@@H]4 CC[C@H]([C@H](C)CCCC(C)C)[C@@]4(C)CC[C@@H]3 [C@@]2(C)C1 |
| IUPAC Name | (5 R,8 R,9 S,10 S,13 R,14 S,17 R)-2,2,4,4-tetradeuterio-10,13-dimethyl-17-[(2 R)-6-methylheptan-2-yl]-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthrene |
| Molecular Weight (g/mol) | 376.695 |
| Formula Weight | 376.401 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction