missing translation for 'onlineSavingsMsg'
Learn More

5alpha-Hydroxytriptolide, TRC

Product Code. 30007045
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007045 10 mg 10mg
30006441 5 mg 5mg
30006230 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007045 Supplier Toronto Research Chemicals Supplier No. TRCH96000010MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583028-68-6
Molecular Formula C20 H24 O7
Molecular Weight (g/mol) 376.4
Synonym (3bR,4aS,5aS,6R,6aR,7aS,7bS,8aS,8bS)-3b,4,4a,6,6a,7a,7b,8b,9,10-Decahydro-3b,6-dihydroxy-8b-methyl-6a-(1-methylethyl)trisoxireno[4b,5:6,7:8a,9]phenanthro[1,2-c]furan-1(3H)-one, (-)-5α-Hydroxytryptolide, (5R)-5-Hydroxytriptolide, LLDT 8, PG 701;
SMILES CC(C)[C@]12O[C@H]1[C@@H]3O[C@@]34[C@@]5(C)CCC6=C(COC6=O)[C@]5(O)C[C@@H]7O[C@]47[C@@H]2O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H24O7/c1-8(2)18-12(26-18)13-20(27-13)16(3)5-4-9-10(7-24-14(9)21)17(16,23)6-11-19(20,25-11)15(18)22/h8,11-13,15,22-23H,4-7H2,1-3H3/t11-,12-,13-,15+,16-,17+,18-,19+,20+/m0/s1
Formula Weight 376.1522
Chemical Name or Material 5alpha-Hydroxytriptolide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.