missing translation for 'onlineSavingsMsg'
Learn More

(5aR,10aR)-Tetrahydrodithiazolo[3,4-a:3',4'-d]pyrazine-5,10(3H,8H)-dione, TRC

Product Code. 30427485
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427485 100 mg
30422600 50 mg
30415014 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427485 Supplier Toronto Research Chemicals Supplier No. TRCT205684100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72744-67-3
Molecular Formula C8H10N2O2S2
Molecular Weight (g/mol) 230.31
Synonym Pidotimod Impurity B
IUPAC Name (5aR,10aR)-1,3,5a,6,8,10a-hexahydrodi[1,3]thiazolo[3,2-b:2',3'-f]pyrazine-5,10-dione
SMILES O=C1[C@@H]2CSCN2C(=O)[C@@H]3CSCN13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H10N2O2S2/c11-7-5-1-13-3-9(5)8(12)6-2-14-4-10(6)7/h5-6H,1-4H2/t5-,6-/m0/s1
Formula Weight 230.0184
Chemical Name or Material (5aR,10aR)-Tetrahydrodithiazolo[3,4-a:3',4'-d]pyrazine-5,10(3H,8H)-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.