Learn More
Enzo Life Sciences 5g:Cycloheximide
Brand:  Enzo Life Sciences ALX-380-269-G005
Encompass_Preferred
5g:Cycloheximide
Product Code. 15119607
  • 268.62 € / 5g
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Cycloheximide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H341-Suspected of causing genetic defects.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.