missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-1,2-Dibromopropane-d6, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | 1,2-Dibromopropane-D6 |
| CAS | 51209-47-3 |
| Molecular Formula | C3 D6 Br2 |
| Quantity | 5 g |
| Synonym | Propane-1,1,1,2,3,3-d6, 2,3-dibromo- (9 CI), Propane-1,1,1,2,3,3-d, 2,3-dibromo- (6 CI), 1,2-Dibromopropane-d6, 1,2-Dibromopropane-D6 |
| InChI Formula | InChI=1 S/C3H6Br2/c1-3(5)2-4/h3H,2H2,1H3/i1D3,2D2,3 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(Br)C([2 H])([2 H])Br |
| IUPAC Name | 1,2-dibromo-1,1,2,3,3,3-hexadeuteriopropane |
| Molecular Weight (g/mol) | 207.9247 |
| Formula Weight | 205.9213 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction