missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromopropane-1,1,1,3,3,3-d6, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | 2-Bromopropane-d6 |
| CAS | 52809-76-4 |
| Molecular Formula | C3 D6 H Br |
| Quantity | 5 g |
| Synonym | 2-Bromopropane-1,1,1,3,3,3-d6, 2-Bromo-1,1,1,3,3,3-hexadeuteropropane |
| InChI Formula | InChI=1 S/C3H7Br/c1-3(2)4/h3H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(Br)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-bromo-1,1,1,3,3,3-hexadeuteriopropane |
| Molecular Weight (g/mol) | 129.03 |
| Formula Weight | 128.0108 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction