missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoyl-d5 Chloride, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Benzoyl-d5 Chloride |
| CAS | 43019-90-5 |
| Molecular Formula | C7 2H5 Cl O |
| Quantity | 5 g |
| InChI Formula | InChI=1 S/C7H5ClO/c8-7(9)6-4-2-1-3-5-6/h1-5 H/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)Cl)c([2 H])c1[2 H] |
| IUPAC Name | 2,3,4,5,6-pentadeuteriobenzoyl chloride |
| Molecular Weight (g/mol) | 145.6 |
| Formula Weight | 145.0343 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction