missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide-d3, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Formamide-D3 |
| CAS | 43380-64-9 |
| Molecular Formula | C D3 N O |
| Quantity | 5 g |
| Synonym | Formamide-d3 (6 CI, 7 CI, 9 CI), Formamide-D3, [2H3]Formamide, Carbamaldehyde-d3, Formimidic acid-d3 |
| InChI Formula | InChI=1 S/CH3NO/c2-1-3/h1H,(H2,2,3)/i1D/hD2 |
| SMILES | [2 H]N([2 H])C(=O)[2 H] |
| IUPAC Name | N,N,1-trideuterioformamide |
| Molecular Weight (g/mol) | 48.0591 |
| Formula Weight | 48.0403 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction