missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1'N-Benzyl Biotin and 3'N-Benzyl Biotin (mixture of regioisomers), TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | 1'N-Benzyl Biotin and 3'N-Benzyl Biotin (mixture of regioisomers) |
| CAS | 2382724-55-0 |
| Molecular Formula | C17H22N2O3S |
| Quantity | 5 mg |
| Synonym | Biotin EP Impurity E (Mixture of Isomers) |
| InChI Formula | InChI=1S/C17H22N2O3S/c20-15(21)9-5-4-8-14-16-13(11-23-14)19(17(22)18-16)10-12-6-2-1-3-7-12/h1-3,6-7,13-14,16H,4-5,8-11H2,(H,18,22)(H,20,21)/t13-,14?,16-/m0/s1 |
| SMILES | O=C1N[C@@]2([H])[C@@](CSC2CCCCC(O)=O)([H])N1CC3=CC=CC=C3 |
| IUPAC Name | 5-[(3aR,6aS)-3-benzyl-2-oxo-3a,4,6,6a-tetrahydro-1H-thieno[3,4-d]imidazol-6-yl]pentanoic acid |
| Molecular Weight (g/mol) | 334.43 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction