missing translation for 'onlineSavingsMsg'
Learn More

AGK2, MedChemExpress

Product Code. 16425827
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16425827

Brand: Medchem Express HY100578

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AGK2 is a selective SIRT2 inhibitor with an IC50 of 3.5 μM. AGK2 inhibits SIRT1 and SIRT3 with IC50s of 30 and 91 μM, respectively.

AGK2 is a selective SIRT2 inhibitor with an IC50 of 3.5 μM. AGK2 inhibits SIRT1 and SIRT3 with IC50s of 30 and 91 μM, respectively.

Specifications

Chemical Name or Material AGK2
CAS 304896-28-4
Color Green Yellow
Purity Grade Notes Research
Molecular Formula C23H13Cl2N3O2
Quantity 5 mg
Solubility Information DMSO : 12.5 mg/mL (28.78 mM; Need ultrasonic)
SMILES O=C(NC1=C2C=CC=NC2=CC=C1)/C(C#N)=C/C3=CC=C(C4=CC(Cl)=CC=C4Cl)O3
Molecular Weight (g/mol) 434.27
Formula Weight 434.27
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.