missing translation for 'onlineSavingsMsg'
Learn More

Amino Tadalafil, TRC

Product Code. 17911605
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911605 5 mg 5mg
30133366 10 mg 10mg
17951175 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17911605 Supplier Toronto Research Chemicals Supplier No. TRCA6295505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 385769-84-6
Molecular Formula C21 H18 N4 O4
Molecular Weight (g/mol) 390.39
Synonym (6R,12aR)-2-Amino-6-(1,3-benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, Aminotadalafil
IUPAC Name (2R,8R)-6-amino-2-(1,3-benzodioxol-5-yl)-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
SMILES NN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H18N4O4/c22-24-9-18(26)25-15(21(24)27)8-13-12-3-1-2-4-14(12)23-19(13)20(25)11-5-6-16-17(7-11)29-10-28-16/h1-7,15,20,23H,8-10,22H2/t15-,20-/m1/s1
Formula Weight 390.1328
Percent Purity >95%
Chemical Name or Material Aminotadalafil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.