missing translation for 'onlineSavingsMsg'
Learn More

Bimiralisib, TRC

Product Code. 30329365
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30329365

Brand: Toronto Research Chemicals TRCB3868405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bimiralisib
CAS 1225037-39-7
Molecular Formula C17 H20 F3 N7 O2
Quantity 5 mg
Synonym 5-(4,6-Di-4-Morpholinyl-1,3,5-triazin-2-yl)-4-(trifluoromethyl)-2-pyridinamine
InChI Formula InChI=1S/C17H20F3N7O2/c18-17(19,20)12-9-13(21)22-10-11(12)14-23-15(26-1-5-28-6-2-26)25-16(24-14)27-3-7-29-8-4-27/h9-10H,1-8H2,(H2,21,22)
SMILES Nc1cc(c(cn1)c2nc(nc(n2)N3CCOCC3)N4CCOCC4)C(F)(F)F
IUPAC Name 5-(4,6-dimorpholin-4-yl-1,3,5-triazin-2-yl)-4-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol) 411.382
Formula Weight 411.163
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.