missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl-D9 Paraben, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Butyl-d9 Paraben |
| CAS | 1216904-65-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C11H5D9O3 |
| Quantity | 5 mg |
| Synonym | 4-Hydroxybenzoic Acid Butyl-d9 Ester, 4-(Butoxy-d9-carbonyl)phenol, 4-Hydroxybenzoic Acid Butyl-d9 Ester, Aseptoform Butyl-d9, Butoben-d9, Butyl-d9 4-Hydroxybenzoate, Butyl-d9 Butex, Butyl-d9 Tegosept-d9, Butyl-d9 Chemosept, n-Butylparaben-d9, Preserval B-d9, SPF-d9, Solbrol B-d9 |
| InChI Formula | InChI=1S/C11H14O3/c1-2-3-8-14-11(13)9-4-6-10(12)7-5-9/h4-7,12H,2-3,8H2,1H3/i1D3,2D2,3D2,8D2 |
| SMILES | OC1=CC=C(C(OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)C=C1 |
| IUPAC Name | 1,1,2,2,3,3,4,4,4-nonadeuteriobutyl 4-hydroxybenzoate |
| Molecular Weight (g/mol) | 203.28 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction