missing translation for 'onlineSavingsMsg'
Learn More

Mericitabine, MedChemExpress

Product Code. 16426467
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16426467

Brand: Medchem Express HY102405mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mericitabine (RG 7128; R-7128) is a nucleoside inhibitor of the HCV NS5B polymerase that acts as an RNA chain terminator and prevents elongation of RNA transcripts during replication.

Mericitabine (RG 7128

Specifications

Chemical Name or Material Mericitabine
CAS 940908-79-2
Color White
Purity Grade Notes Research
Molecular Formula C18H26FN3O6
Quantity 5 mg
Synonym RG 7128 R-7128 PSI 6130 diisobutyrate
Solubility Information DMSO : 100 mg/mL (250.37 mM; Need ultrasonic)
SMILES O=C1N=C(C=CN1[C@@H]([C@@](C)([C@@H]2OC(C(C)C)=O)F)O[C@@H]2COC(C(C)C)=O)N
Molecular Weight (g/mol) 399.41
Formula Weight 399.41
Percent Purity 99.47%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.