missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-S-(2-carboxypropyl)-L-cysteine Bis(dicyclohexylammonium) Salt(Mixture of Diastereomers), TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-S-(2-carboxypropyl)-L-cysteine Bis(dicyclohexylammonium) Salt(Mixture of Diastereomers) |
| Molecular Formula | C9H15NO5S . 2(C12H23N) |
| Quantity | 5 mg |
| Synonym | N-Acetyl-3-(2-carboxypropyl)thio]alanine Bis(dicyclohexylammonium) Salt |
| InChI Formula | InChI=1S/2C12H23N.C9H15NO5S/c2*1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5(8(12)13)3-16-4-7(9(14)15)10-6(2)11/h2*11-13H,1-10H2;5,7H,3-4H2,1-2H3,(H,10,11)(H,12,13)(H,14,15)/t;;5?,7-/m..0/s1 |
| SMILES | CC(CSC[C@H](NC(=O)C)C(=O)O)C(=O)O.C1CCC(CC1)NC2CCCCC2.C3CCC(CC3)NC4CCCCC4 |
| IUPAC Name | 3-[(2R)-2-acetamido-2-carboxy-ethyl]sulfanyl-2-methyl-propanoic acid;N-cyclohexylcyclohexanamine |
| Molecular Weight (g/mol) | 611.91954 |
| Formula Weight | 611.4331 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction