missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 5mL Polystyrene Immuno-Tubes

Product Code. 13097573
Change view
Click to view available options
Surface:
High-binding
Medium-binding
Includes:
Star Bottom
Without Star Bottom
Unit Size:
Pack of 2000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Includes Unit Size
13097573 High-binding Star Bottom Pack of 2000
13067573 Medium-binding Without Star Bottom Pack of 2000
13077573 High-binding Without Star Bottom Pack of 2000
13087573 Medium-binding Star Bottom Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13097573 Supplier Greiner Bio-One Supplier No. 115071
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for determining hormone levels.

  • Round bottom
  • Dia. x Height: 12 x 75mm
  • Available with either medium or high binding capacities
  • With or without star at bottom
  • Star at the bottom of the tubes serve to increase the surface and thus make it possible to bind amounts of antigens or antibodies
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Determining hormone levels
Material Polystyrene
Product Type Immuno Tube
Surface High-binding
Length (Metric) 75 mm
Diameter (Metric) Outer 12 mm
Volume (Metric) 5 mL
Includes Star Bottom
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.