missing translation for 'onlineSavingsMsg'
Learn More

(5R)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-5-[[(methylsulfonyl)oxy]methyl]-2-oxazolidinone, TRC

Product Code. 30132393
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132393 25 mg 25mg
30011174 100 mg 100mg
30010046 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132393 Supplier Toronto Research Chemicals Supplier No. TRCF59497025MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174649-09-3
Molecular Formula C15 H19 F N2 O6 S
Molecular Weight (g/mol) 374.38
Synonym (R)-[3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl]methyl methanesulfonate, (5R)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-5-[[(methylsulfonyl)oxy]methyl]-2-oxazolidinone, 2-Oxazolidinone, 3-[3-fluoro-4-(4-morpholinyl)phenyl]-5-[[(methylsulfonyl)oxy]methyl]-, (5R)-, 2-Oxazolidinone, 3-[3-fluoro-4-(4-morpholinyl)phenyl]-5-[[(methylsulfonyl)oxy]methyl]-, (R)-, (R)-[3-(3-Fluoro-4-morpholin-4-ylphenyl)-2-oxo-5-oxazolidinyl]methyl methanesulfonate, Linezolid Related Compound D
IUPAC Name [(5R)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl methanesulfonate
SMILES CS(=O)(=O)OC[C@H]1CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H19FN2O6S/c1-25(20,21)23-10-12-9-18(15(19)24-12)11-2-3-14(13(16)8-11)17-4-6-22-7-5-17/h2-3,8,12H,4-7,9-10H2,1H3/t12-/m1/s1
Formula Weight 374.0948
Chemical Name or Material (5R)-[3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl]methyl Methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.