missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (5Z)-7-Oxozeaenol
GREENER_CHOICE

Product Code. 18725187
Change view
Click to view available options
:
1mg
Product Code.
18725187 1mg
1 options
This item is not returnable. View return policy

Product Code. 18725187

Brand: Tocris Bioscience™ 3604/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective TAK1 MAPKKK inhibitor

(5Z)-7-Oxozeaenol is a resorcyclic lactone of fungal origin that acts as a potent and selective transforming growth factor-β-activated kinase 1 (TAK1) mitogen-activated protein kinase kinase kinase (MAPKKK) inhibitor (IC50 = 8 nM). Displays > 33-fold and > 62-fold selectivity over MEKK1 and MEKK4 respectively. Inhibits IL-1-induced activation of NF-κB (IC50 = 83 nM) and JNK/p38. Inhibits production of inflammatory mediators, and sensitizes cells to TRAIL- and TNF-α-induced apoptosis in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type (5Z)-7-Oxozeaenol
Purity >99%
Molecular Weight (g/mol) 362.37
Inhibitors (5Z)-7-Oxozeaenol

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.