missing translation for 'onlineSavingsMsg'
Learn More

(5Z)-rel-Cloprostenol Sodium, TRC

Product Code. 18072830
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
Unit Size:
10mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18072830 10 mg 10mg
18062540 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18072830 Supplier Toronto Research Chemicals Supplier No. TRCC58732510MG
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55028-72-3
Molecular Formula C22 H28 Cl O6 . Na
Molecular Weight (g/mol) 446.9
Synonym 5-Heptenoic acid, 7-[(1R,2R,3R,5S)-2-[(1E,3R)-4-(3-chlorophenoxy)-3-hydroxy-1-buten-1-yl]-3,5-dihydroxycyclopentyl]-, sodium salt (1:1), (5Z)-rel-, 5-Heptenoic acid, 7-[(1R,2R,3R,5S)-2-[(1E,3R)-4-(3-chlorophenoxy)-3-hydroxy-1-butenyl]-3,5-dihydroxycyclopentyl]-, monosodium salt, (5Z)-rel- (9CI), 5-Heptenoic acid, 7-[2-[4-(3-chlorophenoxy)-3-hydroxy-1-butenyl]-3,5-dihydroxycyclopentyl]-, monosodium salt, [1α(Z),2β(1E,3R*),3α,5α]-(±)-, 5-Heptenoic acid, 7-[2-[4-(3-chlorophenoxy)-3-hydroxy-1-butenyl]-3,5-dihydroxycyclopentyl]-, monosodium salt, [1α(Z),2β(1E,3R*),3α,5α]-, Ciosin, Cloprostenol sodium salt, Estrumat, Estrumate, ICI 80996, Planate, Vetmate, Cloprostenol Sodium
IUPAC Name sodium;(Z)-7-[(1R,2R,3R,5S)-2-[(E,3R)-4-(3-chlorophenoxy)-3-hydroxybut-1-enyl]-3,5-dihydroxycyclopentyl]hept-5-enoate
SMILES [Na+].O[C@@H](COc1cccc(Cl)c1)\C=C\[C@H]2[C@H](O)C[C@H](O)[C@@H]2C\C=C/CCCC(=O)[O-]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H29ClO6.Na/c23-15-6-5-7-17(12-15)29-14-16(24)10-11-19-18(20(25)13-21(19)26)8-3-1-2-4-9-22(27)28;/h1,3,5-7,10-12,16,18-21,24-26H,2,4,8-9,13-14H2,(H,27,28);/q;+1/p-1/b3-1-,11-10+;/t16-,18-,19-,20+,21-;/m1./s1
Formula Weight 446.1472
Percent Purity >95%
Chemical Name or Material Cloprostenol Sodium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.